Anti-Nitroso-D,L-Citrulline Rat Polyclonal Antibody (Unconjugated)

Lieferant: US Biological
C5806-01D
USBIC5806-01DEA 739 EUR
USBIC5806-01D
Anti-Nitroso-D,L-Citrulline Rat Polyclonal Antibody (Unconjugated)
Antikörper
Type: Primary
Antigen: Nitroso-D,L-Citrulline
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR