Anti-Nitroso-D,L-Citrulline Rabbit Polyclonal Antibody (Unconjugated)
Lieferant: US Biological
C5806-01E
USBIC5806-01EEA 739 EUR
USBIC5806-01E
Anti-Nitroso-D,L-Citrulline Rabbit Polyclonal Antibody (Unconjugated)
Antikörper
Type: Primary
Antigen: Nitroso-D,L-Citrulline
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen