Anti-Citrulline Rabbit polyclonal antibody
Lieferant: US Biological
C5806-02
USBIC5806-02EA 843 EUR
USBIC5806-02
Anti-Citrulline Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Citrulline
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human