Anti-D,L-Citrulline Rabbit Polyclonal Antibody (Unconjugated)
Lieferant: US Biological
C5806-50
USBIC5806-50EA 473 EUR
USBIC5806-50
Anti-D,L-Citrulline Rabbit Polyclonal Antibody (Unconjugated)
Antikörper
Type: Primary
Antigen: D,L-Citrulline
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen