Anti-JUN Rabbit Polyclonal Antibody

Lieferant: US Biological
C5815-14
USBIC5815-14EA 602 EUR
USBIC5815-14
Anti-JUN Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunoblot: A 1:1000

Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Avian, Bovine, Human, Mouse, Pig, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR