Anti-Clostridium sp. Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
C5853-25B
USBIC5853-25BEA 459 EUR
USBIC5853-25B
Anti-Clostridium sp. Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: Immunofluorescence: 1:500

Type: Primary
Antigen: Clostridium sp.
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR