Anti-COL5 Goat Polyclonal Antibody (Biotin)

Lieferant: US Biological
C7510-55W
USBIC7510-55WEA 754 EUR
USBIC7510-55W
Anti-COL5 Goat Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: Immunochemistry: 1:20-1:40; ELISA: 1:1000-1:4000

Type: Primary
Antigen: COL5 (collagen, type V)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR