Anti-Colon Carcinoma Mouse Monoclonal Antibody [clone: CC-3]

Lieferant: US Biological
C7554-50B
USBIC7554-50BEA 530 EUR
USBIC7554-50B
Anti-Colon Carcinoma Mouse Monoclonal Antibody [clone: CC-3]
Antikörper
Recommended dilution: Immunochemistry: 1:10 Stains acetone or ethanol fixed tissues

Type: Primary
Antigen: Colon Carcinoma
Clonality: Monoclonal
Clone: CC-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR