Anti-CFI Mouse monoclonal antibody [clone: 3D6]

Lieferant: US Biological
C7850-90
USBIC7850-90EA 735 EUR
USBIC7850-90
Anti-CFI Mouse monoclonal antibody [clone: 3D6]
Antikörper
Type: Primary
Antigen: CFI (complement factor I)
Clonality: Monoclonal
Clone: 3D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Lambda
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR