Anti-Cotinine 3 Rabbit Polyclonal Antibody

Lieferant: US Biological
C7904-50B
USBIC7904-50BEA 1170 EUR
USBIC7904-50B
Anti-Cotinine 3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Cotinine 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR