Anti-Cotinine 4 Rabbit Polyclonal Antibody
Lieferant: US Biological
C7904-50D
USBIC7904-50DEA 1170 EUR
USBIC7904-50D
Anti-Cotinine 4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Cotinine 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen