Anti-Cotinine Mouse Monoclonal Antibody [clone: 11H39]

Lieferant: US Biological
C7904-51E
USBIC7904-51EEA 742 EUR
USBIC7904-51E
Anti-Cotinine Mouse Monoclonal Antibody [clone: 11H39]
Antikörper
Type: Primary
Antigen: Cotinine
Clonality: Monoclonal
Clone: 11H39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR