Anti-CAMP CT Rabbit Polyclonal Antibody

Lieferant: US Biological
C7906-74
USBIC7906-74EA 754 EUR
USBIC7906-74
Anti-CAMP CT Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CAMP CT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR