Anti-C-Reactive Protein Mouse Monoclonal Antibody (Biotin) [clone: 1.BB.101]
Lieferant: US Biological
C7907-04P
USBIC7907-04PEA 1490 EUR
USBIC7907-04P
Anti-C-Reactive Protein Mouse Monoclonal Antibody (Biotin) [clone: 1.BB.101]
Antikörper
Type: Primary
Antigen: C-Reactive Protein
Clonality: Monoclonal
Clone: 1.BB.101
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human