Anti-Creatine Kinase BB Isoenzyme Mouse monoclonal antibody [clone: 10B2593]

Lieferant: US Biological
C7910-14E
USBIC7910-14EEA 482 EUR
USBIC7910-14E
Anti-Creatine Kinase BB Isoenzyme Mouse monoclonal antibody [clone: 10B2593]
Antikörper
Type: Primary
Antigen: Creatine Kinase BB Isoenzyme
Clonality: Monoclonal
Clone: 10B2593
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR