Anti-CREB Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
C7915-05H-BIOT
USBIC7915-05H-BIOTEA 884 EUR
USBIC7915-05H-BIOT
Anti-CREB Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CREB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR