Anti-Cryptosporidium sp. Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B2304]
Lieferant: US Biological
C7944-22B
USBIC7944-22BEA 607 EUR
USBIC7944-22B
Anti-Cryptosporidium sp. Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B2304]
Antikörper
Recommended dilution: Flow Cytometry: 1:5-1:50;
Type: Primary
Antigen: Cryptosporidium sp.
Clonality: Monoclonal
Clone: 10B2304
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen