Anti-ACKR3 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
C8376-25E-BIOT
USBIC8376-25E-BIOTEA 884 EUR
USBIC8376-25E-BIOT
Anti-ACKR3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ACKR3 (atypical chemokine receptor 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse