Anti-CBS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: US Biological
C8948-50C-HRP
USBIC8948-50C-HRPEA 884 EUR
USBIC8948-50C-HRP
Anti-CBS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: CBS (cystathionine-beta-synthase)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse