Anti-CBS Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
C8948-50C-PE
USBIC8948-50C-PEEA 884 EUR
USBIC8948-50C-PE
Anti-CBS Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: CBS (cystathionine-beta-synthase)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse