Anti-CYP4X1 Rabbit Polyclonal Antibody

Lieferant: US Biological
C9095-45X
USBIC9095-45XEA 667 EUR
USBIC9095-45X
Anti-CYP4X1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: CYP4X1 (cytochrome P450, family 4, subfamily X, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR