Anti-Cytokeratin 18, Multiepitope Cocktail Mouse Monoclonal Antibody [clone: 5F223]

Lieferant: US Biological
C9097-23M
USBIC9097-23MEA 854 EUR
USBIC9097-23M
Anti-Cytokeratin 18, Multiepitope Cocktail Mouse Monoclonal Antibody [clone: 5F223]
Antikörper
Type: Primary
Antigen: Cytokeratin 18, Multiepitope Cocktail
Clonality: Monoclonal
Clone: 5F223
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Hamster, Human, Mammalian, Mouse, Pig, Rat, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR