Anti-DAF-3 Rabbit polyclonal antibody

Lieferant: US Biological
D1021-01A
USBID1021-01AEA 614 EUR
USBID1021-01A
Anti-DAF-3 Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: DAF-3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: C. elegans
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR