Anti-Dcr-1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
D1499-08A-BIOT
USBID1499-08A-BIOTEA 884 EUR
USBID1499-08A-BIOT
Anti-Dcr-1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Dcr-1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: C. elegans
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR