Anti-RYBP Rabbit Polyclonal Antibody
Lieferant: US Biological
D1882-01J
USBID1882-01JEA 470 EUR
USBID1882-01J
Anti-RYBP Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml
Type: Primary
Antigen: RYBP (RING1 and YY1 binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat