Anti-Delta 4 Rabbit polyclonal antibody

Lieferant: US Biological
D2728-01
USBID2728-01EA 607 EUR
USBID2728-01
Anti-Delta 4 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: Delta 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR