Anti-Delta 4 Rabbit polyclonal antibody

Lieferant: US Biological
D2728-01D
USBID2728-01DEA 613 EUR
USBID2728-01D
Anti-Delta 4 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Immunochemistry: 5-15 ?g/ml; ELISA: 1:3000-1:15000; Western Blot: 1:300-1:1500

Type: Primary
Antigen: Delta 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR