Anti-Deoxyuridine Triphosphate Nucleotidohydrolase Rabbit polyclonal antibody

Lieferant: US Biological
D3206-25A
USBID3206-25AEA 561 EUR
USBID3206-25A
Anti-Deoxyuridine Triphosphate Nucleotidohydrolase Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: Deoxyuridine Triphosphate Nucleotidohydrolase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR