Anti-Dexamethasone Sheep polyclonal antibody

Lieferant: US Biological
D3221-95A
USBID3221-95AEA 776 EUR
USBID3221-95A
Anti-Dexamethasone Sheep polyclonal antibody
Antikörper
Recommended dilution: ELISA: 0.5 ?g/ml

Type: Primary
Antigen: Dexamethasone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR