Anti-Dexamethasone Hemisuccinate Rabbit polyclonal antibody

Lieferant: US Biological
D3221-96H
USBID3221-96HEA 540 EUR
USBID3221-96H
Anti-Dexamethasone Hemisuccinate Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: Dexamethasone Hemisuccinate
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR