Anti-Diamine Oxidase Rabbit Polyclonal Antibody

Lieferant: US Biological
D3375-02-100
USBID3375-02-100EA 838 EUR
USBID3375-02-100
Anti-Diamine Oxidase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Diamine Oxidase
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR