Anti-Dkk-2 CT Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
D3810-03B-AP
USBID3810-03B-APEA 884 EUR
USBID3810-03B-AP
Anti-Dkk-2 CT Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: Dkk-2 CT
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human