Anti-Digoxigenin Sheep polyclonal antibody

Lieferant: US Biological
D8029-50C
USBID8029-50CEA 614 EUR
USBID8029-50C
Anti-Digoxigenin Sheep polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:100-1:5000

Type: Tag
Antigen: Digoxigenin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR