Anti-Digoxigenin Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: US Biological
D8029-50H2
USBID8029-50H2EA 372 EUR
USBID8029-50H2
Anti-Digoxigenin Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:5000; Western Blot: 1:500
Type: Tag
Antigen: Digoxigenin
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen