Anti-Digoxin Mouse monoclonal antibody [clone: 10B2747]

Lieferant: US Biological
D8030-11D
USBID8030-11DEA 572 EUR
USBID8030-11D
Anti-Digoxin Mouse monoclonal antibody [clone: 10B2747]
Antikörper
Type: Primary
Antigen: Digoxin
Clonality: monoclonal
Clone: 10B2747
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR