Anti-Digoxin Mouse monoclonal antibody [clone: 9L158]

Lieferant: US Biological
D8030-14D
USBID8030-14DEA 742 EUR
USBID8030-14D
Anti-Digoxin Mouse monoclonal antibody [clone: 9L158]
Antikörper
Type: Primary
Antigen: Digoxin
Clonality: monoclonal
Clone: 9L158
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR