Anti-Digoxin Mouse monoclonal antibody [clone: 9L159]

Lieferant: US Biological
D8030-14E
USBID8030-14EEA 742 EUR
USBID8030-14E
Anti-Digoxin Mouse monoclonal antibody [clone: 9L159]
Antikörper
Type: Primary
Antigen: Digoxin
Clonality: monoclonal
Clone: 9L159
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR