Anti-Digoxin Goat polyclonal antibody

Lieferant: US Biological
D8030-22A
USBID8030-22AEA 368 EUR
USBID8030-22A
Anti-Digoxin Goat polyclonal antibody
Antikörper
Type: Primary
Antigen: Digoxin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR