Anti-DNP Rabbit Polyclonal Antibody
Lieferant: US Biological
D8060-09-100
USBID8060-09-100EA 742 EUR
USBID8060-09-100
Anti-DNP Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.1-1.0 ?g/ml; Western Blot: 0.5-5 ?g/ml
Type: Primary
Antigen: DNP (Dinitrophenol)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen