Anti-Dinitrophenol Mouse Monoclonal Antibody [clone: 7C25]
Lieferant: US Biological
D8060-10-100
USBID8060-10-100EA 776 EUR
USBID8060-10-100
Anti-Dinitrophenol Mouse Monoclonal Antibody [clone: 7C25]
Antikörper
Recommended dilution: ELISA: 0.1-1 ?g; Western Blot: 0.5-5 ?g/ml
Type: Primary
Antigen: Dinitrophenol
Clonality: Monoclonal
Clone: 7C25
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgE
Reactivity:
Bestellen