Anti-Dopamine Transporter Rat Monoclonal Antibody [clone: 10B2323]

Lieferant: US Biological
D9020-01J
USBID9020-01JEA 863 EUR
USBID9020-01J
Anti-Dopamine Transporter Rat Monoclonal Antibody [clone: 10B2323]
Antikörper
Recommended dilution: Immunochemistry: 1:1000-1:10000

Type: Primary
Antigen: Dopamine Transporter
Clonality: monoclonal
Clone: 10B2323
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR