Anti-Dopamine Transporter Rabbit Polyclonal Antibody

Lieferant: US Biological
D9020-19A
USBID9020-19AEA 775 EUR
USBID9020-19A
Anti-Dopamine Transporter Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:1000

Type: Primary
Antigen: Dopamine Transporter
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR