Anti-DUSP14 Rabbit Polyclonal Antibody

Lieferant: US Biological
D9840-48F
USBID9840-48FEA 667 EUR
USBID9840-48F
Anti-DUSP14 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: DUSP14 (dual specificity phosphatase 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR