Anti-DUSP15 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
D9840-49A-PE
USBID9840-49A-PEEA 884 EUR
USBID9840-49A-PE
Anti-DUSP15 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: DUSP15 (dual specificity phosphatase 15)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR