Anti-Ebola Virus Mouse Monoclonal Antibody [clone: 3G309]
Lieferant: US Biological
E1003-500
USBIE1003-500EA 532 EUR
USBIE1003-500
Anti-Ebola Virus Mouse Monoclonal Antibody [clone: 3G309]
Antikörper
Type: Primary
Antigen: Ebola Virus
Clonality: monoclonal
Clone: 3G309
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen