Anti-ELMO1 Rabbit polyclonal antibody

Lieferant: US Biological
E2248-02A
USBIE2248-02AEA 649 EUR
USBIE2248-02A
Anti-ELMO1 Rabbit polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: ELMO1 (engulfment and cell motility 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR