Anti-ELMO1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
E2248-02A-FITC
USBIE2248-02A-FITCEA 884 EUR
USBIE2248-02A-FITC
Anti-ELMO1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: ELMO1 (engulfment and cell motility 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR