Anti-ELMO1 Goat polyclonal antibody
Lieferant: US Biological
E2248-02D
USBIE2248-02DEA 601 EUR
USBIE2248-02D
Anti-ELMO1 Goat polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 1-2?g/ml
Type: Primary
Antigen: ELMO1 (engulfment and cell motility 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat, Zebrafish