Anti-EMD Rabbit polyclonal antibody

Lieferant: US Biological
E2257-01B
USBIE2257-01BEA 608 EUR
USBIE2257-01B
Anti-EMD Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: EMD (emerin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR