Anti-EMD Rabbit polyclonal antibody

Lieferant: US Biological
E2257-02
USBIE2257-02EA 633 EUR
USBIE2257-02
Anti-EMD Rabbit polyclonal antibody
Antikörper
Recommended dilution: Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: EMD (emerin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR